Learn More
Meso Scale Discovery V-PLEX Human IL-13 Kit
SDP
Catalog No. 5000081202
Encompass
The Meso Scale Discovery V-PLEX Human IL-13 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 13 (IL-13) in various biological fluids It includes a 96-well plate with a monoclonal antibody for IL-13 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay detects 0 03-2500 pg/mL and takes about 3 5 hours It uses MSD s SECTOR Imager for high sensitivity The kit provides reagents for one 96-well plate allowing 40 samples in duplicate
Catalog No. 50-000-81202 Supplier Meso Scale Discovery Supplier No. K151ODD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-13 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 13 (IL-13) in various biological fluids It includes a 96-well plate with a monoclonal antibody for IL-13 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay detects 0 03-2500 pg/mL and takes about 3 5 hours It uses MSD s SECTOR Imager for high sensitivity The kit provides reagents for one 96-well plate allowing 40 samples in duplicate