Learn More
Meso Scale Discovery V-PLEX Human IL-15 Kit
SDP
Catalog No. 5000078841
Encompass
The Meso Scale Discovery V-PLEX Human IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-15 a detection antibody conjugated to an electrochemiluminescent label calibrators and diluents The assay has a sensitivity of 0 23 pg/mL and a broad dynamic range from 0 49-1500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantification of IL-15 levels in human samples
Catalog No. 50-000-78841 Supplier Meso Scale Discovery Supplier No. K151RDD2
The Meso Scale Discovery V-PLEX Human IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 15 (IL-15) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-15 a detection antibody conjugated to an electrochemiluminescent label calibrators and diluents The assay has a sensitivity of 0 23 pg/mL and a broad dynamic range from 0 49-1500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantification of IL-15 levels in human samples