Learn More
Meso Scale Discovery V-PLEX Human IL-16 Kit
SDP
Catalog No. 5000080547
Encompass
The V-PLEX Human IL-16 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human interleukin 16 (IL-16) in various biological fluids This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive IL-16 quantitation with a detection range of 0 05-1000 pg/mL and a minimum detectable dose of 0 02 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high throughput screening
Catalog No. 50-000-80547 Supplier Meso Scale Discovery Supplier No. K151RED2
The V-PLEX Human IL-16 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human interleukin 16 (IL-16) in various biological fluids This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive IL-16 quantitation with a detection range of 0 05-1000 pg/mL and a minimum detectable dose of 0 02 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high throughput screening