Learn More
Meso Scale Discovery V-PLEX Human IL-16 Kit
SDP
Catalog No. 5000082200
Encompass
The V-PLEX Human IL-16 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 16 (IL-16) in serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-82200 Supplier Meso Scale Discovery Supplier No. K151RED1
The V-PLEX Human IL-16 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 16 (IL-16) in serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete after a 2 hour incubation