Learn More
Meso Scale Discovery V-PLEX Human IL-17A/F Kit, Single-analyte assay, 96-well plate format
SDP
Catalog No. NC2682016
Encompass
The MESO SCALE DISCOVERY V-PLEX Human IL-17A/F Kit is an analytic tool designed to precisely measure human IL-17A/F. This kit utilizes electrochemiluminescence (ECL) detection technology for accurate and reproducible results, making it ideal for scientific research into inflammatory responses and immune system regulation.
  • High performance with exceptional sensitivity for reliable biomarker screening.
  • Consistent results across different lots promote data integrity.
  • Utilizes an analytically validated platform known for quality in cytokine detection.
  • Single-analyte format allows focused research without multiplexing complexities.
  • Compatible with various biological specimens, including serum, plasma, and urine.
Catalog No. NC2682016 Supplier Meso Scale Discovery Supplier No. K151WND2
The MESO SCALE DISCOVERY V-PLEX Human IL-17A/F Kit is an analytic tool designed to precisely measure human IL-17A/F. This kit utilizes electrochemiluminescence (ECL) detection technology for accurate and reproducible results, making it ideal for scientific research into inflammatory responses and immune system regulation.
  • High performance with exceptional sensitivity for reliable biomarker screening.
  • Consistent results across different lots promote data integrity.
  • Utilizes an analytically validated platform known for quality in cytokine detection.
  • Single-analyte format allows focused research without multiplexing complexities.
  • Compatible with various biological specimens, including serum, plasma, and urine.