Learn More
Meso Scale Discovery V-PLEX Human IL-17A/F Kit
SDP
Catalog No. 5000081902
Encompass
The V-PLEX Human IL-17A/F Kit is a multiplex immunoassay that allows for the simultaneous measurement of human interleukin 17A (IL-17A) and interleukin 17F (IL-17F) in cell culture supernatants serum and plasma This assay utilizes electrochemiluminescence (ECL) technology on the MSD platform for high sensitivity and broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to quantitatively determine IL-17A and IL-17F concentrations This multiplex assay allows efficient quantification of both analytes from a single small sample volume
Catalog No. 50-000-81902 Supplier Meso Scale Discovery Supplier No. K151WND1
The V-PLEX Human IL-17A/F Kit is a multiplex immunoassay that allows for the simultaneous measurement of human interleukin 17A (IL-17A) and interleukin 17F (IL-17F) in cell culture supernatants serum and plasma This assay utilizes electrochemiluminescence (ECL) technology on the MSD platform for high sensitivity and broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators and controls needed to quantitatively determine IL-17A and IL-17F concentrations This multiplex assay allows efficient quantification of both analytes from a single small sample volume