Learn More
Meso Scale Discovery V-PLEX Human IL-17B Kit
SDP
Catalog No. 5000081481
Encompass
The V-PLEX Human IL-17B Kit from Meso Scale Discovery is a sensitive immunoassay using electrochemiluminescence to measure IL-17B in serum plasma and other fluids It includes a 96-well plate with a monoclonal antibody for IL-17B a detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a lyophilized calibrator Sensitivity is 0 3 pg/mL with a quantification limit of 2500 pg/mL The assay takes about 3 5 hours for 96 tests aiding in IL-17B level determination in research
Catalog No. 50-000-81481 Supplier Meso Scale Discovery Supplier No. K151WOD1
The V-PLEX Human IL-17B Kit from Meso Scale Discovery is a sensitive immunoassay using electrochemiluminescence to measure IL-17B in serum plasma and other fluids It includes a 96-well plate with a monoclonal antibody for IL-17B a detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a lyophilized calibrator Sensitivity is 0 3 pg/mL with a quantification limit of 2500 pg/mL The assay takes about 3 5 hours for 96 tests aiding in IL-17B level determination in research