Learn More
Meso Scale Discovery V-PLEX Human IL-17C Kit
SDP
Catalog No. 5000081830
Encompass
The Meso Scale Discovery V-PLEX Human IL-17C Kit is used to quantitatively measure human interleukin 17C (IL-17C) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to complete the assay Results can be read on MSD s SECTOR Imager instruments with a typical lower limit of detection of 0 05 pg/mL
Catalog No. 50-000-81830 Supplier Meso Scale Discovery Supplier No. K151WPD1
The Meso Scale Discovery V-PLEX Human IL-17C Kit is used to quantitatively measure human interleukin 17C (IL-17C) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium calibrators and reagents required to complete the assay Results can be read on MSD s SECTOR Imager instruments with a typical lower limit of detection of 0 05 pg/mL