Learn More
Meso Scale Discovery V-PLEX Human IL-17D Kit
SDP
Catalog No. 5000081483
Encompass
The V-PLEX Human IL-17D Kit from Meso Scale Discovery is a sandwich immunoassay that allows for the quantitative measurement of human interleukin 17D (IL-17D) in cell culture supernatants serum plasma and other biological fluids The assay utilizes electrochemiluminescence for high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 hours to complete and has a sensitivity of 0 3 pg/mL with an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81483 Supplier Meso Scale Discovery Supplier No. K151WQD1
Add to Cart
Add to Cart
The V-PLEX Human IL-17D Kit from Meso Scale Discovery is a sandwich immunoassay that allows for the quantitative measurement of human interleukin 17D (IL-17D) in cell culture supernatants serum plasma and other biological fluids The assay utilizes electrochemiluminescence for high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 hours to complete and has a sensitivity of 0 3 pg/mL with an upper limit of quantification of 2500 pg/mL