Learn More
Meso Scale Discovery V-PLEX Human IL-17D Kit
SDP
Catalog No. 5000081896
Encompass
The V-PLEX Human IL-17D Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human interleukin 17D (IL-17D) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantitation of IL-17D with a detection range of 0 05-1000 pg/mL and a minimum detectable dose of 0 03 pg/mL The kit includes a 96-well plate detection antibody calibrators controls diluent read buffer and plate seals enabling accurate measurement of IL-17D for immunology and inflammation research
Catalog No. 50-000-81896 Supplier Meso Scale Discovery Supplier No. K151WQD2
The V-PLEX Human IL-17D Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human interleukin 17D (IL-17D) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantitation of IL-17D with a detection range of 0 05-1000 pg/mL and a minimum detectable dose of 0 03 pg/mL The kit includes a 96-well plate detection antibody calibrators controls diluent read buffer and plate seals enabling accurate measurement of IL-17D for immunology and inflammation research