Learn More
Meso Scale Discovery V-PLEX Human IL-1a Kit
SDP
Catalog No. 5000079224
Encompass
The Meso Scale Discovery V-PLEX Human IL-1alpha Kit is a highly sensitive immunoassay that uses electrochemiluminescent detection technology for the quantitative measurement of human interleukin 1 alpha (IL-1alpha) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-1alpha a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 0 06-1500 pg/mL and exhibits no cross-reactivity with other cytokines The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79224 Supplier Meso Scale Discovery Supplier No. K151RBD2
The Meso Scale Discovery V-PLEX Human IL-1alpha Kit is a highly sensitive immunoassay that uses electrochemiluminescent detection technology for the quantitative measurement of human interleukin 1 alpha (IL-1alpha) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for IL-1alpha a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 0 06-1500 pg/mL and exhibits no cross-reactivity with other cytokines The assay takes approximately 3 5 hours to complete