Learn More
Meso Scale Discovery V-PLEX Human IL-1a Kit
SDP
Catalog No. 5000082197
Encompass
The Meso Scale Discovery V-PLEX Human IL-1alpha Kit is a sensitive immunoassay for measuring human interleukin 1 alpha (IL-1alpha) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager detecting IL-1alpha concentrations as low as 0 04 pg/mL
Catalog No. 50-000-82197 Supplier Meso Scale Discovery Supplier No. K151RBD1
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Human IL-1alpha Kit is a sensitive immunoassay for measuring human interleukin 1 alpha (IL-1alpha) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager detecting IL-1alpha concentrations as low as 0 04 pg/mL