Learn More
Meso Scale Discovery V-PLEX Human IL-1beta Kit, 1 plate
SDP
Catalog No. NC1847242
Encompass
Human interleukin-1beta (IL-1beta), also known as IL-1F2, is a 30.7 kDa pro-inflammatory cytokine that is produced by activated macrophages. IL-1beta stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity.
Catalog No. NC1847242 Supplier Meso Scale Discovery Supplier No. K151QPD1
Human interleukin-1beta (IL-1beta), also known as IL-1F2, is a 30.7 kDa pro-inflammatory cytokine that is produced by activated macrophages. IL-1beta stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity.