Learn More
Meso Scale Discovery V-PLEX Human IL-2 Kit, 5 plates
SDP
Catalog No. NC1243018
Encompass
The Human IL-2 Kit provides the assay-specific components for the quantitative determination of endogenous IL-2 in human urine, serum, plasma, and cell culture supernatants.
Catalog No. NC1243018 Supplier Meso Scale Discovery Supplier No. K151QQD2
Add to Cart
Add to Cart
The Human IL-2 Kit provides the assay-specific components for the quantitative determination of endogenous IL-2 in human urine, serum, plasma, and cell culture supernatants.