Learn More
Meso Scale Discovery V-PLEX Human IL-3 Kit
SDP
Catalog No. 5000081485
Encompass
The Meso Scale Discovery V-PLEX Human IL-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a monoclonal antibody to human IL-3 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 08 pg/mL and a 7-log dynamic range from 0 08-600 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 tests in duplicate
Catalog No. 50-000-81485 Supplier Meso Scale Discovery Supplier No. K151WZD1
The Meso Scale Discovery V-PLEX Human IL-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a monoclonal antibody to human IL-3 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 08 pg/mL and a 7-log dynamic range from 0 08-600 pg/mL Sample incubation time is 2 hours The kit can perform up to 94 tests in duplicate