Learn More
Meso Scale Discovery V-PLEX Human IL-3 Kit
SDP
Catalog No. 5000081920
Encompass
The V-PLEX Human IL-3 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody specific for human IL-3 a detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-1500 pg/mL It requires 2 5 hours total assay time This ECL immunoassay allows sensitive accurate and rapid quantification of human IL-3
This item is not returnable. View return policy
Catalog No. 50-000-81920 Supplier Meso Scale Discovery Supplier No. K151WZD2
The V-PLEX Human IL-3 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody specific for human IL-3 a detection antibody conjugated to an ECL label calibrators and buffers for sample dilution The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-1500 pg/mL It requires 2 5 hours total assay time This ECL immunoassay allows sensitive accurate and rapid quantification of human IL-3