Learn More
Meso Scale Discovery V-PLEX Human IL-4 Kit
SDP
Catalog No. 5000078791
Encompass
The Meso Scale Discovery V-PLEX Human IL-4 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It includes a 96-well plate with a capture antibody for IL-4 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay detects 0 03-2500 pg/mL and takes about 3 5 hours It uses MSD s SECTOR Imager for high sensitivity The kit provides reagents for one 96-well plate allowing 40 samples in duplicate
Catalog No. 50-000-78791 Supplier Meso Scale Discovery Supplier No. K151QRD2
The Meso Scale Discovery V-PLEX Human IL-4 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It includes a 96-well plate with a capture antibody for IL-4 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay detects 0 03-2500 pg/mL and takes about 3 5 hours It uses MSD s SECTOR Imager for high sensitivity The kit provides reagents for one 96-well plate allowing 40 samples in duplicate