Learn More
Meso Scale Discovery V-PLEX Human IL-4 Kit
SDP
Catalog No. 5000079184
Encompass
The Meso Scale Discovery V-PLEX Human IL-4 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It uses MSD s V-PLEX technology for high sensitivity and a wide dynamic range The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators for a standard curve It detects IL-4 from 0 10 pg/mL to 10 000 pg/mL with a total assay time of 3 5 hours
Catalog No. 50-000-79184 Supplier Meso Scale Discovery Supplier No. K151QRD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-4 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It uses MSD s V-PLEX technology for high sensitivity and a wide dynamic range The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators for a standard curve It detects IL-4 from 0 10 pg/mL to 10 000 pg/mL with a total assay time of 3 5 hours