Learn More
Meso Scale Discovery V-PLEX Human IL-5 Kit, Single-analyte assay, 96-well plate format, Sensitivity 0.14 pg/mL, Dynamic range 0.14 - 562 pg/mL, Human IL-5 analyte, Serum, Plasma, Urine, Cell Culture samples
SDP
Catalog No. NC2018786
Encompass
The MESO SCALE DISCOVERY V-PLEX Human IL-5 Kit is a high-performance, analytically validated immunoassay designed for the quantitative detection of human Interleukin-5 (IL-5). This robust kit offers accurate and reproducible results with excellent lot-to-lot consistency, making it ideal for cytokine and chemokine research in immunology and inflammation studies.
  • Utilizes a 96-well plate format
  • Quantitative detection of human IL-5
  • High sensitivity measurement with a LLOD of 0.14 pg/mL
  • Dynamic range of 0.14 - 562 pg/mL
  • Compatible with diverse biological samples: Serum, Plasma, Urine, Cell Culture Supernatant
  • Mouse Monoclonal capture antibody
  • Rat Monoclonal detection antibody
  • Suitable for research use only
  • Compatible with various MSD SECTOR and QuickPlex instruments
Catalog No. NC2018786 Supplier Meso Scale Discovery Supplier No. K151QSD2
The MESO SCALE DISCOVERY V-PLEX Human IL-5 Kit is a high-performance, analytically validated immunoassay designed for the quantitative detection of human Interleukin-5 (IL-5). This robust kit offers accurate and reproducible results with excellent lot-to-lot consistency, making it ideal for cytokine and chemokine research in immunology and inflammation studies.
  • Utilizes a 96-well plate format
  • Quantitative detection of human IL-5
  • High sensitivity measurement with a LLOD of 0.14 pg/mL
  • Dynamic range of 0.14 - 562 pg/mL
  • Compatible with diverse biological samples: Serum, Plasma, Urine, Cell Culture Supernatant
  • Mouse Monoclonal capture antibody
  • Rat Monoclonal detection antibody
  • Suitable for research use only
  • Compatible with various MSD SECTOR and QuickPlex instruments