Learn More
Meso Scale Discovery V-PLEX Human IL-6 Kit
SDP
Catalog No. 5000082323
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 6 (IL-6) in various biological fluids It includes a 96-well plate with a capture antibody for IL-6 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay detects IL-6 in the range of 0 2-2500 pg/mL and takes about 3 5 hours It uses an instrument like the MSD QuickPlex SQ 120 to measure emitted light providing sensitive and accurate IL-6 quantitation
Catalog No. 50-000-82323 Supplier Meso Scale Discovery Supplier No. K151QXD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-6 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 6 (IL-6) in various biological fluids It includes a 96-well plate with a capture antibody for IL-6 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay detects IL-6 in the range of 0 2-2500 pg/mL and takes about 3 5 hours It uses an instrument like the MSD QuickPlex SQ 120 to measure emitted light providing sensitive and accurate IL-6 quantitation