Learn More
Meso Scale Discovery V-PLEX Human IL-7 Kit
SDP
Catalog No. 5000080143
Encompass
The Meso Scale Discovery V-PLEX Human IL-7 Kit is a highly sensitive and quantitative immunoassay for the detection of human interleukin 7 (IL-7) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence technology and provides accurate reproducible results in just 3 5 hours The V-PLEX platform has a lower limit of detection of 0 10 pg/mL and a broad dynamic range of up to 4 logs This assay requires a minimal sample volume of 50 L and is designed for use on Meso Scale Discovery instruments The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator
Catalog No. 50-000-80143 Supplier Meso Scale Discovery Supplier No. K151RCD2
The Meso Scale Discovery V-PLEX Human IL-7 Kit is a highly sensitive and quantitative immunoassay for the detection of human interleukin 7 (IL-7) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence technology and provides accurate reproducible results in just 3 5 hours The V-PLEX platform has a lower limit of detection of 0 10 pg/mL and a broad dynamic range of up to 4 logs This assay requires a minimal sample volume of 50 L and is designed for use on Meso Scale Discovery instruments The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator