Learn More
Meso Scale Discovery V-PLEX Human IL-8 Kit
SDP
Catalog No. 5000079223
Encompass
The V-PLEX Human IL-8 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) to measure human interleukin 8 (IL-8) in various biological fluids It includes a 96-well plate with a monoclonal antibody to IL-8 a detection antibody with an ECL label calibrators and buffers The assay s sensitivity is 0 09 pg/mL with a range of 0 09-1500 pg/mL and takes 2 5 hours The ECL platform ensures high sensitivity wide range and minimal interference making it a reliable tool for IL-8 research and inflammation studies
Catalog No. 50-000-79223 Supplier Meso Scale Discovery Supplier No. K151RAD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human IL-8 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) to measure human interleukin 8 (IL-8) in various biological fluids It includes a 96-well plate with a monoclonal antibody to IL-8 a detection antibody with an ECL label calibrators and buffers The assay s sensitivity is 0 09 pg/mL with a range of 0 09-1500 pg/mL and takes 2 5 hours The ECL platform ensures high sensitivity wide range and minimal interference making it a reliable tool for IL-8 research and inflammation studies