Learn More
Meso Scale Discovery V-PLEX Human IL-9 Kit
SDP
Catalog No. 5000078720
Encompass
The Meso Scale Discovery V-PLEX Human IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 9 (IL-9) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody to human IL-9 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of human IL-9
Catalog No. 50-000-78720 Supplier Meso Scale Discovery Supplier No. K151XCD1
The Meso Scale Discovery V-PLEX Human IL-9 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 9 (IL-9) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody to human IL-9 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of human IL-9