Learn More
Meso Scale Discovery V-PLEX Human Insulin Kit
SDP
Catalog No. 5000080544
Encompass
The Meso Scale Discovery V-PLEX Human Insulin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human insulin in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 50-25 000 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit includes sufficient reagents for one 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-80544 Supplier Meso Scale Discovery Supplier No. K151S5D1
The Meso Scale Discovery V-PLEX Human Insulin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human insulin in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 50-25 000 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit includes sufficient reagents for one 96-well plate and can assay 40 samples in duplicate