Learn More
Meso Scale Discovery V-PLEX Human Insulin Kit
SDP
Catalog No. 5000081683
Encompass
The V-PLEX Human Insulin Kit from Meso Scale Discovery is a multiplex immunoassay for measuring insulin in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators controls buffers and wash solutions The assay detects 50-25 000 pg/mL with high sensitivity specificity and reproducibility using MSD s electrochemiluminescent technology It tests 40 samples in duplicate
Catalog No. 50-000-81683 Supplier Meso Scale Discovery Supplier No. K151S5D2
Add to Cart
Add to Cart
The V-PLEX Human Insulin Kit from Meso Scale Discovery is a multiplex immunoassay for measuring insulin in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators controls buffers and wash solutions The assay detects 50-25 000 pg/mL with high sensitivity specificity and reproducibility using MSD s electrochemiluminescent technology It tests 40 samples in duplicate