Learn More
Meso Scale Discovery V-PLEX HUMAN IP-10 GEN B KIT, Singleplex assay, 96-well plate, Analyte: IP-10, Dynamic Range: 0.12 - 1,240 pg/mL
SDP
Catalog No. NC2827028
Encompass
V-PLEX HUMAN IP-10 GEN B KIT, Singleplex assay, 96-well plate, Analyte: IP-10, Dynamic Range: 0.12 - 1,240 pg/mL
Catalog No. NC2827028 Supplier Meso Scale Discovery Supplier No. K151ANLD2
V-PLEX HUMAN IP-10 GEN B KIT, Singleplex assay, 96-well plate, Analyte: IP-10, Dynamic Range: 0.12 - 1,240 pg/mL