Learn More
Meso Scale Discovery V-PLEX Human Leptin Kit
SDP
Catalog No. 5000080424
Encompass
The Meso Scale Discovery V-PLEX Human Leptin Kit is an electrochemiluminescent immunoassay for measuring human leptin in serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for rapid sensitive detection with a detection limit of 26 pg/mL and quantification up to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody conjugated to ruthenium calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range ensuring accurate leptin quantification
Catalog No. 50-000-80424 Supplier Meso Scale Discovery Supplier No. K151V5D2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Human Leptin Kit is an electrochemiluminescent immunoassay for measuring human leptin in serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for rapid sensitive detection with a detection limit of 26 pg/mL and quantification up to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody conjugated to ruthenium calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range ensuring accurate leptin quantification