Learn More
Meso Scale Discovery V-PLEX Human MCP-1 Kit, 1 plate, single-analyte, 96-well format, sensitivity 0.09 pg/mL, dynamic range 0.09 - 375 pg/mL, for serum, plasma, urine, cell culture supernatant
SDP
Catalog No. NC2820017
Encompass
V-PLEX Human MCP-1 Kit, 1 plate, single-analyte, 96-well format, sensitivity 0.09 pg/mL, dynamic range 0.09 - 375 pg/mL, for serum, plasma, urine, cell culture supernatant
Catalog No. NC2820017 Supplier Meso Scale Discovery Supplier No. K151NND1
V-PLEX Human MCP-1 Kit, 1 plate, single-analyte, 96-well format, sensitivity 0.09 pg/mL, dynamic range 0.09 - 375 pg/mL, for serum, plasma, urine, cell culture supernatant