Learn More
Meso Scale Discovery V-PLEX Human MCP-4 Kit
SDP
Catalog No. 5000080767
Encompass
The V-PLEX Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the cytokine MCP-4 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The assay has a detection range of 0 08-500 pg/mL and takes approximately 3 5 hours to complete It utilizes electrochemiluminescence for high sensitivity and requires a Meso Scale Discovery instrument for analysis The kit provides reagents for one 96-well plate which can test up to 40 samples in duplicate
Catalog No. 50-000-80767 Supplier Meso Scale Discovery Supplier No. K151NOD1
The V-PLEX Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the cytokine MCP-4 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The assay has a detection range of 0 08-500 pg/mL and takes approximately 3 5 hours to complete It utilizes electrochemiluminescence for high sensitivity and requires a Meso Scale Discovery instrument for analysis The kit provides reagents for one 96-well plate which can test up to 40 samples in duplicate