Learn More
Meso Scale Discovery V-PLEX Human MCP-4 Kit
SDP
Catalog No. 5000081184
Encompass
The V-PLEX Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 4 (MCP-4) across multiple samples using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for measuring MCP-4 in serum plasma and cell culture supernatants
Catalog No. 50-000-81184 Supplier Meso Scale Discovery Supplier No. K151NOD2
The V-PLEX Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 4 (MCP-4) across multiple samples using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for measuring MCP-4 in serum plasma and cell culture supernatants