Learn More
Meso Scale Discovery V-PLEX Human MCP-4 Kit
SDP
Catalog No. 5000082267
Encompass
The V-PLEX Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the concentration of human monocyte chemoattractant protein 4 (MCP-4) in serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator The assay has a typical lower limit of detection of 0 03 pg/mL and an upper limit of 10 000 pg/mL for MCP-4
Catalog No. 50-000-82267 Supplier Meso Scale Discovery Supplier No. K151NOD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the concentration of human monocyte chemoattractant protein 4 (MCP-4) in serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator The assay has a typical lower limit of detection of 0 03 pg/mL and an upper limit of 10 000 pg/mL for MCP-4