Learn More
Meso Scale Discovery V-PLEX Human MDC Kit
SDP
Catalog No. 5000079924
Encompass
The Meso Scale Discovery V-PLEX Human MDC Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage-Derived Chemokine (MDC) in human serum plasma and cell culture supernatants The assay has high sensitivity wide dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79924 Supplier Meso Scale Discovery Supplier No. K151NPD1
The Meso Scale Discovery V-PLEX Human MDC Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage-Derived Chemokine (MDC) in human serum plasma and cell culture supernatants The assay has high sensitivity wide dynamic range and requires as little as 25 L of sample The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay takes approximately 3 5 hours to complete