Learn More
Meso Scale Discovery V-PLEX Human MDC Kit
SDP
Catalog No. 5000080765
Encompass
The V-PLEX Human MDC Kit from Meso Scale Discovery is a multiplex immunoassay that measures the levels of human MDC (CCL22) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for human MDC along with detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescent assay has a sensitivity of 1 3 pg/mL and a range of 1 3-1000 pg/mL The kit allows for the quantitative measurement of MDC levels with high sensitivity and wide dynamic range
Catalog No. 50-000-80765 Supplier Meso Scale Discovery Supplier No. K151NPD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human MDC Kit from Meso Scale Discovery is a multiplex immunoassay that measures the levels of human MDC (CCL22) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for human MDC along with detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The electrochemiluminescent assay has a sensitivity of 1 3 pg/mL and a range of 1 3-1000 pg/mL The kit allows for the quantitative measurement of MDC levels with high sensitivity and wide dynamic range