Learn More
Meso Scale Discovery V-PLEX Human PP Kit
SDP
Catalog No. 5000079697
Encompass
The V-PLEX Human PP Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of Pancreatic Polypeptide (PP) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The electrochemiluminescent assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 1500 pg/mL The total assay time is 3 5 hours This kit can be used for research purposes to study PP levels in relation to pancreatic function and metabolism
Catalog No. 50-000-79697 Supplier Meso Scale Discovery Supplier No. K151Y5D2
The V-PLEX Human PP Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of Pancreatic Polypeptide (PP) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The electrochemiluminescent assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 1500 pg/mL The total assay time is 3 5 hours This kit can be used for research purposes to study PP levels in relation to pancreatic function and metabolism