Learn More
Meso Scale Discovery V-PLEX Human Proinflammatory Panel I (4-Plex), 5 plates
SDP
Catalog No. NC1243017
Encompass
The Human Proinflammatory Panel I (4-Plex) Kit provides assay-specific components for the quantitative determination of endogenous IFN-gamma, IL-1beta, IL-6, and TNF-alpha in human urine, serum, plasma, and cell culture supernatants.
Catalog No. NC1243017 Supplier Meso Scale Discovery Supplier No. K15052D2
The Human Proinflammatory Panel I (4-Plex) Kit provides assay-specific components for the quantitative determination of endogenous IFN-gamma, IL-1beta, IL-6, and TNF-alpha in human urine, serum, plasma, and cell culture supernatants.