Learn More
Meso Scale Discovery V-PLEX Human Tie-2 Kit
SDP
Catalog No. 5000079996
Encompass
The Meso Scale Discovery V-PLEX Human TIE-2 Kit is a multiplex immunoassay for measuring human TIE-2 in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed with MSD s MESO QuickPlex SQ 120 instrument This kit ensures accurate measurement of human TIE-2 levels
Catalog No. 50-000-79996 Supplier Meso Scale Discovery Supplier No. K151KWD1
The Meso Scale Discovery V-PLEX Human TIE-2 Kit is a multiplex immunoassay for measuring human TIE-2 in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed with MSD s MESO QuickPlex SQ 120 instrument This kit ensures accurate measurement of human TIE-2 levels