Learn More
Meso Scale Discovery V-PLEX Human Tie-2 Kit
SDP
Catalog No. 5000080845
Encompass
The Meso Scale Discovery V-PLEX Human TIE-2 Kit is a multiplex immunoassay for measuring human TIE-2 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification with just 25 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an optimized assay diluent It quantifies TIE-2 over 1 5-1000 pg/mL with high specificity and minimal cross-reactivity
Catalog No. 50-000-80845 Supplier Meso Scale Discovery Supplier No. K151KWD2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Human TIE-2 Kit is a multiplex immunoassay for measuring human TIE-2 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification with just 25 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an optimized assay diluent It quantifies TIE-2 over 1 5-1000 pg/mL with high specificity and minimal cross-reactivity