Learn More
Meso Scale Discovery V-PLEX Human Tie-2 Kit
SDP
Catalog No. 5000082236
Encompass
The Meso Scale Discovery V-PLEX Human TIE-2 Kit is a multiplex immunoassay for measuring human TIE-2 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate TIE-2 quantification with a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s platforms This product supports angiogenesis research and drug discovery by reliably measuring TIE-2 levels
Catalog No. 50-000-82236 Supplier Meso Scale Discovery Supplier No. K151KWD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human TIE-2 Kit is a multiplex immunoassay for measuring human TIE-2 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate TIE-2 quantification with a broad dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s platforms This product supports angiogenesis research and drug discovery by reliably measuring TIE-2 levels