Learn More
Meso Scale Discovery V-PLEX Human TSLP Kit 25 PL
SDP
Catalog No. 5000079701
Encompass
The Meso Scale Discovery V-PLEX Human TSLP Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human thymic stromal lymphopoietin (TSLP) in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with a capture antibody specific for human TSLP a detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a sensitivity of 0 31 pg/mL and a broad dynamic range from 0 31-2000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantification of human TSLP levels
Catalog No. 50-000-79701 Supplier Meso Scale Discovery Supplier No. K151XED4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human TSLP Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human thymic stromal lymphopoietin (TSLP) in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with a capture antibody specific for human TSLP a detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a sensitivity of 0 31 pg/mL and a broad dynamic range from 0 31-2000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantification of human TSLP levels