Learn More
Meso Scale Discovery V-PLEX Human TSLP Kit
SDP
Catalog No. 5000081488
Encompass
The V-PLEX Human TSLP Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of thymic stromal lymphopoietin (TSLP) in human serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81488 Supplier Meso Scale Discovery Supplier No. K151XED2
The V-PLEX Human TSLP Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of thymic stromal lymphopoietin (TSLP) in human serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete