Learn More
Meso Scale Discovery V-PLEX Human TSLP Kit
SDP
Catalog No. 5000081798
Encompass
The V-PLEX Human TSLP Kit is an enzyme-linked immunosorbent assay (ELISA) kit for the quantitative determination of thymic stromal lymphopoietin (TSLP) concentrations in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for human TSLP standards detection antibodies streptavidin-HRP substrate solution and wash and sample diluents The assay has a sensitivity of 16 4 pg/mL and can detect TSLP in the range of 31 3-2000 pg/mL Sample incubation time is 3 5 hours The kit includes sufficient reagents for one 96-well plate
Catalog No. 50-000-81798 Supplier Meso Scale Discovery Supplier No. K151XED1
The V-PLEX Human TSLP Kit is an enzyme-linked immunosorbent assay (ELISA) kit for the quantitative determination of thymic stromal lymphopoietin (TSLP) concentrations in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a monoclonal antibody specific for human TSLP standards detection antibodies streptavidin-HRP substrate solution and wash and sample diluents The assay has a sensitivity of 16 4 pg/mL and can detect TSLP in the range of 31 3-2000 pg/mL Sample incubation time is 3 5 hours The kit includes sufficient reagents for one 96-well plate