Learn More
Meso Scale Discovery V-PLEX Human VCAM-1 Kit
SDP
Supplier:  Meso Scale Discovery K151SRD2
Encompass
The Meso Scale Discovery V-PLEX Human VCAM-1 Kit is a highly sensitive immunoassay that quantitatively measures human VCAM-1 in cell culture supernatants serum plasma and other biological fluids The kit uses electrochemiluminescence detection technology and provides accurate reproducible results in just 3 5 hours The V-PLEX assay requires as little as 25 L of sample and has a lower limit of detection of 0 2 ng/mL This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of lyophilized VCAM-1 calibrator
Catalog No. 50-000-78515