Learn More
Meso Scale Discovery V-PLEX Human VCAM-1 Kit
SDP
Catalog No. 5000078755
Encompass
The V-PLEX Human VCAM-1 Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying human VCAM-1 in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a lyophilized calibrator The detection range is 1 53-2500 pg/mL with minimal cross-reactivity For research use only it enables high-throughput sensitive detection of human VCAM-1
Catalog No. 50-000-78755 Supplier Meso Scale Discovery Supplier No. K151SRD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human VCAM-1 Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying human VCAM-1 in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a lyophilized calibrator The detection range is 1 53-2500 pg/mL with minimal cross-reactivity For research use only it enables high-throughput sensitive detection of human VCAM-1