Learn More
Meso Scale Discovery V-PLEX Human VEGF-C Kit
SDP
Catalog No. 5000078674
Encompass
The V-PLEX Human VEGF-C Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying human VEGF-C in cell culture supernatants serum plasma and other fluids It includes a 96-well plate with capture antibodies for VEGF-C detection antibodies with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a VEGF-C calibrator for a standard curve The assay detects 1 53-2500 pg/mL and shows minimal cross-reactivity with other proteins offering a sensitive and reproducible method for measuring VEGF-C in samples
Catalog No. 50-000-78674 Supplier Meso Scale Discovery Supplier No. K151LTD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human VEGF-C Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying human VEGF-C in cell culture supernatants serum plasma and other fluids It includes a 96-well plate with capture antibodies for VEGF-C detection antibodies with an electrochemiluminescent label calibrator diluent wash buffer read buffer and a VEGF-C calibrator for a standard curve The assay detects 1 53-2500 pg/mL and shows minimal cross-reactivity with other proteins offering a sensitive and reproducible method for measuring VEGF-C in samples