Learn More
Meso Scale Discovery V-PLEX Human VEGF-C Kit
SDP
Catalog No. 5000079158
Encompass
The V-PLEX Human VEGF-C Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human VEGF-C in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against VEGF-C detection antibodies conjugated to an electrochemiluminescent label calibrator diluent wash buffer read buffer and a vial of VEGF-C calibrator to generate a standard curve The assay has a detection range of 1 53-2500 pg/mL and exhibits minimal cross-reactivity with other analytes The kit offers high sensitivity and requires only 25 L of sample
Catalog No. 50-000-79158 Supplier Meso Scale Discovery Supplier No. K151LTD2
The V-PLEX Human VEGF-C Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human VEGF-C in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against VEGF-C detection antibodies conjugated to an electrochemiluminescent label calibrator diluent wash buffer read buffer and a vial of VEGF-C calibrator to generate a standard curve The assay has a detection range of 1 53-2500 pg/mL and exhibits minimal cross-reactivity with other analytes The kit offers high sensitivity and requires only 25 L of sample