Learn More
Meso Scale Discovery V-PLEX Human VEGF-D Kit
SDP
Catalog No. 5000079291
Encompass
The Meso Scale Discovery V-PLEX Human VEGF-D Kit is a multiplex immunoassay that quantitatively measures human VEGF-D in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of VEGF-D on the MESO QuickPlex SQ 120 instrument The V-PLEX kit includes a VEGF-D calibrator and pre-coated plates to enable accurate quantification of VEGF-D levels With minimal sample volume requirements and a streamlined workflow this kit allows high-throughput profiling of VEGF-D which plays a key role in lymphangiogenesis and cancer metastasis
Catalog No. 50-000-79291 Supplier Meso Scale Discovery Supplier No. K151LUD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human VEGF-D Kit is a multiplex immunoassay that quantitatively measures human VEGF-D in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of VEGF-D on the MESO QuickPlex SQ 120 instrument The V-PLEX kit includes a VEGF-D calibrator and pre-coated plates to enable accurate quantification of VEGF-D levels With minimal sample volume requirements and a streamlined workflow this kit allows high-throughput profiling of VEGF-D which plays a key role in lymphangiogenesis and cancer metastasis