Learn More
Meso Scale Discovery V-PLEX Human VEGF-D Kit
SDP
Catalog No. 5000079157
Encompass
The Meso Scale Discovery V-PLEX Human VEGF-D Kit is a multiplex immunoassay that quantitatively measures human VEGF-D in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of VEGF-D on the MESO QuickPlex SQ 120 instrument The kit includes a VEGF-D calibrator and pre-coated plates with capture antibodies enabling high-throughput analysis of VEGF-D levels with low sample volume requirements This assay provides researchers with a tool to further study the role of VEGF-D in cancer and other diseases
Catalog No. 50-000-79157 Supplier Meso Scale Discovery Supplier No. K151LUD2
The Meso Scale Discovery V-PLEX Human VEGF-D Kit is a multiplex immunoassay that quantitatively measures human VEGF-D in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of VEGF-D on the MESO QuickPlex SQ 120 instrument The kit includes a VEGF-D calibrator and pre-coated plates with capture antibodies enabling high-throughput analysis of VEGF-D levels with low sample volume requirements This assay provides researchers with a tool to further study the role of VEGF-D in cancer and other diseases