Learn More
Meso Scale Discovery V-PLEX Human VEGF-D Kit
SDP
Catalog No. 5000079954
Encompass
The Meso Scale Discovery V-PLEX Human VEGF-D Kit is a multiplex immunoassay that quantitatively measures human VEGF-D in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The V-PLEX kit includes a 96-well plate coated with capture antibodies for VEGF-D detection antibodies conjugated to MSD SULFO-TAG calibrators and an MSD Read Buffer for electrochemiluminescent signal detection on MSD instruments The assay has a detection range of 3 5-10 000 pg/mL and requires only 25 L of sample
Catalog No. 50-000-79954 Supplier Meso Scale Discovery Supplier No. K151LUD1
The Meso Scale Discovery V-PLEX Human VEGF-D Kit is a multiplex immunoassay that quantitatively measures human VEGF-D in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The V-PLEX kit includes a 96-well plate coated with capture antibodies for VEGF-D detection antibodies conjugated to MSD SULFO-TAG calibrators and an MSD Read Buffer for electrochemiluminescent signal detection on MSD instruments The assay has a detection range of 3 5-10 000 pg/mL and requires only 25 L of sample