Learn More
Meso Scale Discovery V-PLEX Mouse IFN-gamma Kit, 5 plates
SDP
Catalog No. NC1202777
Encompass
The Mouse IFN-gamma Kit provides assay-specific components for the quantitative determination of endogenous IFN-gamma in mouse urine, serum, plasma, and cell culture supernatants.
Catalog No. NC1202777 Supplier Meso Scale Discovery Supplier No. K152QOD2
The Mouse IFN-gamma Kit provides assay-specific components for the quantitative determination of endogenous IFN-gamma in mouse urine, serum, plasma, and cell culture supernatants.