Learn More
Meso Scale Discovery V-PLEX Mouse IFN-y Kit
SDP
Catalog No. 5000081383
Encompass
The V-PLEX Mouse IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying mouse interferon gamma (IFN-) in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 71 pg/mL with an upper quantification limit of 2500 pg/mL using MSD s electrochemiluminescent technology for high sensitivity and a small sample volume of 25-50 L
Catalog No. 50-000-81383 Supplier Meso Scale Discovery Supplier No. K152QOD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Mouse IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay for quantifying mouse interferon gamma (IFN-) in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 71 pg/mL with an upper quantification limit of 2500 pg/mL using MSD s electrochemiluminescent technology for high sensitivity and a small sample volume of 25-50 L